EMAIL THIS PAGE TO A FRIEND
Neurogenetics
Mutation in the gene encoding lysosomal acid phosphatase (Acp2) causes cerebellum and skin malformation in mouse.
PMID 15503243
Abstract
We report a novel spontaneous mutation named nax in mice, which exhibit delayed hair appearance and ataxia in a homozygote state. Histological analyses of nax brain revealed an overall impairment of the cerebellar cortex. The classical cortical cytoarchitecture was disrupted, the inner granule cell layer was not obvious, the Purkinje cells were not aligned as a Purkinje cell layer, and Bergmann glias did not span the molecular layer. Furthermore, histological analyses of skin showed that the hair follicles were also abnormal. We mapped the nax locus between marker D2Mit158 and D2Mit100 within a region of 800 kb in the middle of chromosome 2 and identified a missense mutation (Gly244Glu) in Acp2, a lysosomal monoesterase. The Glu244 mutation does not affect the stability of the Acp2 transcript, however it renders the enzyme inactive. Ultrastructural analysis of nax cerebellum showed lysosomal storage bodies in nucleated cells, suggesting progressive degeneration as the underlying mechanism. Identification of Acp2 as the gene mutated in nax mice provides a valuable model system for studying the role of Acp2 in cerebellum and skin homeostasis.