EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Origins of selectivities in proline-catalyzed alpha-aminoxylations.
PMID 15506738
Abstract
The mechanisms, transition structures, regioselectivity, and stereoselectivity of proline-catalyzed alpha-aminoxylation reactions were studied with density functional theory (B3LYP/6-31G(d)). The most favorable transition structure involves the nucleophilic attack of the (E)-proline enamine to the oxygen of the nitrosobenzene with proton transfer from the carboxylic acid to the nitrogen. Enamine attack to the oxygen is favored over the attack on the nitrogen due to the greater basicity of the nitrogen. Previously proposed zwitterionic enaminium pathways are highly disfavored due to the energetic penalty associated with charge separations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N24609
Nitrosobenzene, ≥97%
C6H5NO