EMAIL THIS PAGE TO A FRIEND
Journal of ethnopharmacology
3 Beta-(3,4-dihydroxycinnamoyl)-erythrodiol, a cytotoxic constituent of Zygophyllum geslini collected in the Algerian Sahara.
PMID 15507367
Abstract
3beta-(3,4-Dihydroxycinnamoyl)-erythrodiol was isolated as the cytotoxic constituent of the roots of Zygophyllum geslini.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
09258
Erythrodiol, analytical standard
C30H50O2