EMAIL THIS PAGE TO A FRIEND
Journal of cell science
Platelet-activating factor stimulates cytoplasmic alkalinization and granule acidification in human eosinophils.
PMID 15507482
Abstract
The effects of platelet-activating factor (PAF) and IL-5 on intracellular pH were investigated in human eosinophils. Purified peripheral blood eosinophils were loaded with the ratiometric fluorescent pH indicator BCECF-AM ester. Stimulation of eosinophils with PAF produced time-dependent alkalinization of the cytoplasm from an initial pH of 7.1+/-0.04 to 7.5+/-0.05. A similar alkalinization response was produced by the calcium ionophore, ionomycin and by the calcium ATPase inhibitor, thapsigargin. These compounds as well as PAF produce significant increases in cytoplasmic calcium ([Ca2+]i). In contrast, IL-5 and the protein kinase C (PKC) activator phorbol myristate acetate (PMA) did not produce cytoplasmic alkalinization and had no effect on [Ca2+]i in eosinophils. PAF-stimulated alkalinization was not inhibited under conditions that blocked plasma membrane Na+-H+ exchange, proton channel or plasma membrane H+-ATPase activities. Measurements of intragranule pH with a cell permeant pH indicator (LysoSensor Yellow/Blue DND-160), which partitions into intracellular acidic compartments, revealed that PAF-stimulated cytosolic alkalinization correlated with intragranule acidification. These results suggest that the increase in [Ca2+]i after PAF stimulation activates a H+-ATPase present in the granule membranes, leading to enhanced granule acidification and cytoplasmic alkalinization. We propose that granule acidification is an important step in solubilization of major basic protein crystals, which are stored within the granule core, in preparation for degranulation and release of these proteins.
Related Materials