EMAIL THIS PAGE TO A FRIEND
Plant & cell physiology
Low concentrations of propyzamide and oryzalin alter microtubule dynamics in Arabidopsis epidermal cells.
PMID 15509858
Abstract
Previous studies showed that sub-micromolar concentrations of the microtubule-targeting herbicide propyzamide cause a right-handed helical arrangement of cortical microtubule arrays and left-handed twisting in elongating Arabidopsis epidermal cells. When seedlings were grown in the presence of 1-2 microM propyzamide or 50-100 nM oryzalin, we show that microtubules spent more time in a paused state in which they exhibited little net change in length. The drug treatment also resulted in slower growth and shortening velocities, increased catastrophe, and an overall decrease in microtubule turnover. A reduction in microtubule dynamic turnover may underlie the drug-induced rearrangement of cortical arrays.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
442764
Pronamide (Kerb), analytical standard
C12H11Cl2NO
45645
Propyzamide, PESTANAL®, analytical standard
C12H11Cl2NO