EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Restoration of copper metabolism and rescue of hepatic abnormalities in LEC rats, an animal model of Wilson disease, by expression of human ATP7B gene.
PMID 15511628
Abstract
Hepatic abnormalities in Long-Evans Cinnamon (LEC) rats, an animal model of Wilson disease (WD), were restored by the expression of the human ATP7B cDNA under the control of CAG promoter. Expression of ATP7B transcript and protein in the liver of the transgenic rats resulted in the restoration of biosynthesis of holoceruloplasmin and biliary copper excretion. Meanwhile, transgenic rats showed striking improvements in their hepatic abnormalities, i.e., rescue from fulminant hepatitis, late onset of hepatic cholangiofibrosis, suppression of hepatocellular carcinoma and much improved survival rates. Moreover, dramatic decreases were noted both in the levels of hepatic copper and iron in transgenic rats before the occurrence of hepatitis. These results indicated that the human ATP7B product compensated for the deficiency of the endogenous rattus protein and did function in intrahepatic copper transport by secreting copper into the plasma via incorporation into ceruloplasmin and by the excretion of copper into the bile, and that ATP7B is critical to hepatic dysfunctions in WD. This first successful transgenic rescue has important implications for the gene therapy of WD.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
BCR058
Continuous cast copper (O), BCR® certified Reference Material, rod
Cu
31284
Copper, electrolytic, Analytical Reagent, Reag. Ph. Eur., ≥99.9% (complexometric), wire
Cu
326445
Copper, bars (random sizes), ≥99.999% trace metals basis
Cu
254177
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Cu
326488
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Cu
266744
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Cu
349178
Copper, foil, thickness 0.25 mm, 99.98% trace metals basis
Cu
349208
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Cu
12816
Copper, foil, ≥99.8% (complexometric)
Cu
203122
Copper, powder, 99.999% trace metals basis
Cu
266086
Copper, powder, <425 μm, 99.5% trace metals basis
Cu
207780
Copper, powder, <75 μm, 99%
Cu
326453
Copper, powder (spheroidal), 10-25 μm, 98%
Cu
357456
Copper, powder (dendritic), <45 μm, 99.7% trace metals basis
Cu
365327
Copper, rod, diam. 11 mm, 99.9999% trace metals basis
Cu
223409
Copper, shot, −3-+14 mesh, 99%
Cu
520381
Copper, spherical shot, −20-+30 mesh, ≥99.5% trace metals basis
Cu
349224
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Cu
349216
Copper, wire, diam. 1.0 mm, ≥99.9%
Cu
326410
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Cu
326429
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Cu
61139
Copper, turnings, purum p.a., ≥99.0%
Cu
61141
Copper, purum p.a., ≥99.0%, powder
Cu
IRMM522A
Copper, IRMM® certified Reference Material, 0.1 mm foil
Cu
IRMM522C
Copper, IRMM® certified Reference Material, 0.5 mm wire
Cu
349151
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Cu
311405
Copper, ACS reagent, granular, 10-40 mesh, ≥99.90%
Cu
326437
Copper, rod, diam. 3.1 mm, 99.999%
Cu
767476
Copper, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.95% trace metals basis
Cu
774111
Copper, nanopowder, 40-60 nm particle size (SAXS), ≥99.5% trace metals basis
Cu