EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Modular synthesis of formamidines and their formation of stable organogels.
PMID 15514842
Abstract
An improvement in the practical aspects of formamidine synthesis has resulted in the discovery of a class of compounds which produce organogels in protic solvents, presumably through intermolecular hydrogen bonding and pi-pi stacking interactions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F15803
Formamidine acetate salt, 99%
CH4N2 · C2H4O2
268607
Formamidine hydrochloride, 97%
CH4N2 · HCl