EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
General method for facile intramolecular disulfide formation in synthetic peptides.
PMID 15519585
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
143057
Aldrithiol-4, 98%
C10H8N2S2