EMAIL THIS PAGE TO A FRIEND
Organic letters
Stereochemical control of the Passerini reaction.
PMID 15524450
Abstract
A catalytic asymmetric Passerini reaction using tridentate indan (pybox) Cu(II) Lewis acid complex 4 with substrates capable of bidentate coordination has been achieved. The reaction occurs via ligand-accelerated catalysis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45789
Indan, analytical standard
C9H10
I1804
Indan, 95%
C9H10