EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Lung cellular and molecular physiology
Endothelin-1 decreases endothelial NOS expression and activity through ETA receptor-mediated generation of hydrogen peroxide.
PMID 15531748
Abstract
Similar to infants born with persistent pulmonary hypertension of the newborn (PPHN), there is an increase in circulating endothelin-1 (ET-1) and decreased endothelial nitric oxide synthase (eNOS) gene expression in an ovine model of PPHN. These abnormalities lead to vasoconstriction and vascular remodeling. Our previous studies have demonstrated that reactive oxygen species (ROS) levels are elevated in the pulmonary arteries from PPHN lambs and that ET-1 increases ROS production in pulmonary arterial smooth muscle cells (PASMC) in culture. Thus the objective of this study was to determine whether there was a feedback mechanism between the ET-1-mediated increase in ROS in fetal PASMC (FPASMC) and a decrease in eNOS gene expression in fetal pulmonary arterial endothelial cells (FPAEC). Our results indicate that ET-1 increased H2O2 levels in FPASMC in an endothelin A receptor-dependent fashion. This was observed in both FPASMC monoculture and in cocultures of FPASMC and FPAEC. Conversely, ET-1 decreased H2O2 levels in FPAEC monoculture in an endothelin B receptor-dependent fashion. Furthermore, ET-1 decreased eNOS promoter activity by 40% in FPAEC in coculture with FPASMC. Promoter activity was restored in the presence of catalase. In FPAEC in monoculture treated with 0-100 microM H2O2, 12 microM had no effect on eNOS promoter activity, but it increased eNOS protein levels by 50%. However, at 100 microM, H2O2 decreased eNOS promoter activity and protein levels in FPAEC by 79 and 40%, respectively. These data suggest a role for smooth muscle cell-derived H2O2 in ET-1-mediated downregulation of eNOS expression in children born with PPHN.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E7764
Endothelin 1, ≥97% (HPLC), powder
C109H159N25O32S5