EMAIL THIS PAGE TO A FRIEND
Photodermatology, photoimmunology & photomedicine
Psoralen ultraviolet A therapy in 2004.
PMID 15533240
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
73633
Psoralen, analytical standard
C11H6O3