EMAIL THIS PAGE TO A FRIEND
Journal of travel medicine
Bismuth revisited: an effective way to prevent travelers' diarrhea.
PMID 15541227
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF86561354
Bismuth, foil, not light tested, 50x50mm, thickness 0.025mm, permanent polyester support, 99.97%
Bi
GF85045631
Bismuth, foil, not light tested, 50x50mm, thickness 0.01mm, permanent polyester support, 99.97%
Bi
GF80139438
Bismuth, foil, not light tested, 50x50mm, thickness 0.05mm, permanent polyester support, 99.97%
Bi
GF81875447
Bismuth, rod, 120mm, diameter 10.0mm, cast sticks, 100%
Bi
GF71052432
Bismuth, rod, 140mm, diameter 10.0mm, cast sticks, 100%
Bi
GF60966366
Bismuth, rod, 70mm, diameter 10.0mm, cast sticks, 100%
Bi
GF89918500
Bismuth, foil, 25x25mm, thickness 0.25mm, 100%
Bi
GF71570550
Bismuth, foil, 25x25mm, thickness 0.5mm, 100%
Bi
GF58555581
Bismuth, foil, 25x25mm, thickness 1.0mm, 100%
Bi
GF52383506
Bismuth, foil, 25x25mm, thickness 1.0mm, 99.97%
Bi
GF31001960
Bismuth, foil, 25x25mm, thickness 2.0mm, 100%
Bi
GF39813419
Bismuth, foil, 50x50mm, thickness 0.25mm, 100%
Bi
GF46319340
Bismuth, foil, 50x50mm, thickness 0.5mm, 100%
Bi
GF19448017
Bismuth, foil, 50x50mm, thickness 1.0mm, 100%
Bi
GF97656238
Bismuth, foil, 50x50mm, thickness 1.0mm, 99.97%
Bi
GF75246183
Bismuth, foil, 50x50mm, thickness 2.0mm, 100%
Bi
GF46489135
Bismuth, foil, light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 99.97%
Bi
GF86892441
Bismuth, foil, light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 99.97%
Bi
GF40931546
Bismuth, foil, light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 99.97%
Bi
GF35246831
Bismuth, foil, light tested, 25x25mm, thickness 0.02mm, permanent polyester support, 99.97%
Bi
GF95571995
Bismuth, foil, light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 99.97%
Bi
GF12550935
Bismuth, foil, light tested, 50x50mm, thickness 0.015mm, permanent polyester support, 99.97%
Bi
GF09655263
Bismuth, foil, light tested, 50x50mm, thickness 0.01mm, permanent polyester support, 99.97%
Bi
GF71673060
Bismuth, foil, light tested, 50x50mm, thickness 0.025mm, permanent polyester support, 99.97%
Bi
GF09816307
Bismuth, foil, light tested, 50x50mm, thickness 0.02mm, permanent polyester support, 99.97%
Bi
GF91742328
Bismuth, foil, light tested, 50x50mm, thickness 0.05mm, permanent polyester support, 99.97%
Bi
GF24083692
Bismuth, foil, not light tested, 100x100mm, thickness 0.015mm, permanent polyester support, 99.97%
Bi
GF77293592
Bismuth, foil, not light tested, 100x100mm, thickness 0.01mm, permanent polyester support, 99.97%
Bi
GF93912891
Bismuth, foil, not light tested, 100x100mm, thickness 0.025mm, permanent polyester support, 99.97%
Bi
GF19216087
Bismuth, foil, not light tested, 100x100mm, thickness 0.02mm, permanent polyester support, 99.97%
Bi