EMAIL THIS PAGE TO A FRIEND
Journal of Cellular Biochemistry
Characterization of events associated with apoptosis/anoikis induced by snake venom metalloproteinase BaP1 on human endothelial cells.
PMID 15543558