EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Biofunctionalization of fluorescent rare-earth-doped lanthanum phosphate colloidal nanoparticles.
PMID 15547904
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
587176
Lanthanum(III) phosphate hydrate, ≥99.99% trace metals basis
LaO4P · xH2O