EMAIL THIS PAGE TO A FRIEND
Applied microbiology and biotechnology
Hydroxylation activity of P450 BM-3 mutant F87V towards aromatic compounds and its application to the synthesis of hydroquinone derivatives from phenolic compounds.
PMID 15549292
Abstract
Cytochrome P450 BM-3 from Bacillus megaterium is a fatty acid hydroxylase exhibiting selectivity for long-chain substrates (12-20 carbons). Replacement of Phe87 in P450 BM-3 by Val (F87V) greatly increased its activity towards a variety of aromatic and phenolic compounds. The apparent initial reaction rates of F87V as to benzothiophene, indan, 2,6-dichlorophenol, and 2-(benzyloxy)phenol were 227, 204, 129, and 385 nmol min(-1) nmol(-1) P450, which are 220-, 66-, 99-, and 963-fold those of the wild type, respectively. These results indicate that Phe87 plays a critical role in the control of the substrate specificity of P450 BM-3. Furthermore, F87V catalyzed regioselective hydroxylation at the para position of various phenolic compounds. In particular, F87V showed high activity as to the hydroxylation of 2-(benzyloxy)phenol to 2-(benzyloxy)hydroquinone. With F87V as the catalyst, 0.71 mg ml(-1) 2-(benzyloxy)hydroquinone was produced from 1.0 mg ml(-1) 2-(benzyloxy)phenol in 4 h, with a molar yield of 66%.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
183040
2-(Benzyloxy)phenol, 96%
C13H12O2