EMAIL THIS PAGE TO A FRIEND
Lipids
Reaction of mono-epoxidized conjugated linoleic acid ester with boron trifluoride etherate complex.
PMID 15554159
Abstract
The reaction of methyl 11,12-E-epoxy-9Z-octadecenoate (1) with boron trifluoride etherate furnished a mixture of methyl 12-oxo-10E-octadecenoate (3a) and methyl 11-oxo-9E-octadecenoate (3b) in 66% yield. Methyl 9,10-Z-epoxy-11E-octadecenoate (2) with boron trifluoride etherate furnished a mixture of methyl 9-oxo-10E-octadecenoate (4a, 45%) and methyl 10-oxo-11E-octadecenoate (4b, 19%). A plausible mechanism is proposed for these reactions, which involves the attack on the epoxy ring system by BF3, followed by deprotonation, oxo formation, and double bond migration to give a mixture of two positional alpha,beta-unsaturated C18 enone ester derivatives (3a/3b, 4a/4b). The structures of these C18 enone ester derivatives (3a/3b, 4a/4b) were identified by a combination of NMR spectroscopic and mass spectrometric analyses.