EMAIL THIS PAGE TO A FRIEND
Biophysical chemistry
Aspartame and aspartame derivatives effect human thrombin catalytic activity.
PMID 15572260
Abstract
The study of small Asp-Phe analogs was undertaken since this dipeptide sequence is critical in fibrinogen recognition and catalysis. The inhibition of clotting activity by Asp-Phe-methyl ester (aspartame), formyl-Asp-Phe-methyl ester and acetyl-Asp-Phe was biphasic in all cases, indicating the presence of at least two binding sites. The N-terminally blocked derivatives are stronger inhibitors than aspartame. In contrast, tosyl-Gly-Pro-Arg-p'-nitroanilide hydrolysis was inhibited minimally by Asp-Phe-methyl, ester [Ki(app)=98 mM]. Acetyl-Asp-Phe inhibition of thrombin amidase activity was biphasic, tenfold stronger and appeared to be strongly cooperative. These results are discussed with respect to the inhibition of alpha-thrombin by ATP.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5139
Asp-Phe methyl ester, ≥98%
C14H18N2O5