EMAIL THIS PAGE TO A FRIEND
Acta poloniae pharmaceutica
E- and Z-p-methoxycinnamic acid from Aquilegia vulgaris.
PMID 15575598
Abstract
Two isomers E- and Z-p-methoxycinnamic acid were isolated from the leaves with stems of A. vulgaris L. and their structures were determined by 1HMR and 13C NMR spectral data.
Related Materials