EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
A simple and efficient highly enantioselective synthesis of alpha-ionone and alpha-damascone.
PMID 15575784
Abstract
An efficient highly enantioselective (ee > or =99%) synthesis of alpha-ionone and alpha-damascone is described. Both enantiomers of title compounds were synthesized through two straightforward pathways diverging from enantiopure (R)- or (S)-alpha-cyclogeraniol. These versatile building blocks were obtained by regioselective ZrCl(4)-promoted biomimetic cyclization of (6S)- or (6R)-(Z)-6,7-epoxygeraniol, respectively, followed by deoxygenation of the so formed secondary alcohol. The chiral information was encoded by a highly regioselecive Sharpless asymmetric dihydroxylation of inexpensive geranyl acetate.
Related Materials