EMAIL THIS PAGE TO A FRIEND
Cell and Tissue Research
Immunohistochemical distribution of proteins belonging to the receptor-mediated and the mitochondrial apoptotic pathways in human placenta during gestation.
PMID 15578274