EMAIL THIS PAGE TO A FRIEND
Physiology & behavior
Prolyl-leucyl-glycinamide shares some effects with oxytocin but decreases oxytocin levels.
PMID 15581670
Abstract
Oxytocin treatment in rats induces long-lasting antistress and growth promoting effects. This study investigated whether prolyl-leucyl-glycinamide (PLG) (the c-terminal tripeptide of oxytocin) or tocinoic acid (the ring structure of oxytocin) could induce some of these effects in male rats. For this purpose, PLG (2 or 10 mg/kg, s.c.) or tocinoic acid (1 mg/kg, s.c.) was administered to rats once a day for 3 or 5 days. Blood pressure, heart rate, spontaneous motor activity, nociceptive thresholds, and the survival of ischaemic musculocutaneous flaps were measured. In addition, endogenous oxytocin levels and plasma levels of some hormones known to be influenced by oxytocin were determined. PLG (2 mg/kg, s.c., but not 10 mg/kg, s.c.) decreased diastolic blood pressure (p<0.05) and locomotor activity (p<0.05). PLG (10 mg/kg, s.c.) decreased gastrin (p<0.05) and endogenous oxytocin levels in plasma (p<0.01). Tocinoic acid decreased locomotor activity (p<0.05), but did not affect any of the other parameters measured. In conclusion, this study showed that both PLG and tocinoic acid decrease locomotor activity. In addition, PLG also induced some other effects similar to those induced by oxytocin treatment but when administered in high doses it decreased oxytocin levels.