EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Synthesis and aminoacyl-tRNA synthetase inhibitory activity of aspartyl adenylate analogs.
PMID 15582453
Abstract
Three nonhydrolyzable aspartyl adenylate analogs have been prepared and tested as inhibitors of E. coli aspartyl-tRNA synthetase. 5'-O-[N-(L-Aspartyl)sulfamoyl]adenosine is a potent competitive inhibitor (K(i) = 15 nM) whereas L-aspartol adenylate is a weaker inhibitor (K(i) = 45 microM) with respect to aspartic acid. The corresponding ketomethylphosphonate (a novel isosteric replacement) is also a strong inhibitor (K(i) = 123 nM).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
515876
Imidazole trifluoromethanesulfonate salt, 97%
C3H4N2 · CHF3O3S