EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Direct high-performance liquid chromatographic separation of unusual secondary amino acids and a comparison of the performances of Chirobiotic T and TAG columns.
PMID 15584244
Abstract
Two macrocyclic glycopeptide antibiotic-type chiral stationary phases (CSPs) based on native teicoplanin and teicoplanin aglycone, Chirobiotic T and TAG, respectively, were evaluated with regard to the high-performance liquid chromatographic separation of the enantiomers of 10 secondary alpha-amino acids (imino acids). The chromatographic results are given as the retention, separation and resolution factors, together with the enantioselective free energy difference corresponding to the separation of the enantiomers. By application of these two CSPs, excellent resolutions were achieved for the investigated compounds by using reversed-phase mobile mode systems. The separation conditions were optimized by variation of the mobile phase composition. The difference in enantioselective free energy between the aglycone CSP and the teicoplanin CSP for these particular amino acids ranged between 0.70 and -1.83 kJ mol(-1). It was established that better enantioseparations of the secondary alpha-amino acids were attained in most cases on the aglycone CSP.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
12018AST Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
12019AST Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
12020AST Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
12010AST Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 3 mm
12021AST Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 5 cm × 4.6 mm
12022AST Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
12023AST Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
12024AST Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
12034AST Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm
14018AST Astec® CHIROBIOTIC® TAG Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
14019AST Astec® CHIROBIOTIC® TAG Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
14020AST Astec® CHIROBIOTIC® TAG Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
14022AST Astec® CHIROBIOTIC® TAG Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
14023AST Astec® CHIROBIOTIC® TAG Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
14024AST Astec® CHIROBIOTIC® TAG Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
14232AST Astec® CHIROBIOTIC® TAG Chiral HPLC Column, 16 μm particle size, L × I.D. 15 cm × 10 mm
14034AST Astec® CHIROBIOTIC® TAG Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm
14044AST Astec® CHIROBIOTIC® TAG Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 21.2 mm