EMAIL THIS PAGE TO A FRIEND
Klinicheskaia laboratornaia diagnostika
[Pyrogallic method for the determination of protein in urine (literature survey)].
PMID 15584391
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P8759
Pyrogallol Red
C19H12O8S