EMAIL THIS PAGE TO A FRIEND
Vascular and endovascular surgery
Efficacy of dextran solutions in vascular surgery.
PMID 15592628
Abstract
The purpose of this paper is to discuss the role and efficacy of dextran in vascular procedures using evidence-based data from the review of surgical literature. A medline search using "dextran,'' "vascular surgery,'' and "antiplatelet therapy'' as keywords was performed for English-language articles. Further references were obtained through cross-referencing the bibliography cited in each work. Dextran is commonly used in carotid endarterectomy (CEA) patients where the embolic rate is reduced by 46%, resulting in fewer procedure-related strokes. As a prophylactic agent against thrombosis, multiple randomized studies have reported its benefit over other antithrombotic medications. Dextran is also particularly useful in "difficult'' infragenicular lower extremity bypasses where artificial grafts (such as polytetrafluoroethylene [PTFE] or umbilical vein) are used in the setting of poor outflow vessels, or those with composite grafts and small-caliber venous conduits. Distal bypasses with adjunctive procedures (eg, arteriovenous fistula or anastomotic cuffs) also have a better outcome with the addition of dextran. Dextran has numerous important implications in vascular surgery, in particular with CEA patients or "difficult'' infragenicular bypasses. Its effectiveness with endovascular stents remains unknown.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
00271 Dextran, analytical standard, for GPC, 25,000
00891 Dextran, analytical standard, for GPC, 50,000
00893 Dextran, analytical standard, for GPC, 150,000
00269 Dextran, analytical standard, for GPC, 5,000
00268 Dextran, analytical standard, for GPC, 1,000
31430 Dextran, analytical standard, for GPC, Set Mp 1,000-400,000
00895 Dextran, analytical standard, for GPC, 410,000
00894 Dextran, analytical standard, for GPC, 270,000
00892 Dextran, analytical standard, for GPC, 80,000
00270 Dextran, analytical standard, for GPC, 12,000
00896 Dextran, analytical standard, for GPC, 670,000
31394 Dextran, enzymatic synth.
D0731000 Dextran, European Pharmacopoeia (EP) Reference Standard
D0731005 Dextran 1, European Pharmacopoeia (EP) Reference Standard
D0734000 Dextran 10 for calibration, European Pharmacopoeia (EP) Reference Standard
D0737000 Dextran 250 for calibration, European Pharmacopoeia (EP) Reference Standard
D0733000 Dextran 4 for calibration, European Pharmacopoeia (EP) Reference Standard
D0735000 Dextran 40 for calibration, European Pharmacopoeia (EP) Reference Standard
D0738000 Dextran 40 for performance test, European Pharmacopoeia (EP) Reference Standard
D0739000 Dextran 60/70 for performance test, European Pharmacopoeia (EP) Reference Standard
D0736000 Dextran 70 for calibration, European Pharmacopoeia (EP) Reference Standard
D9260 Dextran from Leuconostoc mesenteroides, average mol wt 9,000-11,000
D5376 Dextran from Leuconostoc mesenteroides, average mol wt 1,500,000-2,800,000
D8821 Dextran from Leuconostoc mesenteroides, average mol wt 60,000-76,000
D1662 Dextran from Leuconostoc mesenteroides, average mol wt 35,000-45,000
D4876 Dextran from Leuconostoc mesenteroides, average mol wt 150,000
31416 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 1,000
31425 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 670,000
31417 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 5,000
31424 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 410,000