EMAIL THIS PAGE TO A FRIEND
Journal of Zhejiang University. Science. B
Separation and identification of cis and trans isomers of 2-butene-1,4-diol and lafutidine by HPLC and LC-MS.
PMID 15593397
Abstract
The cis and trans isomers separation of 2-butene-1,4-diol and lafutidine were studied by HPLC on two kinds of chiral columns: (S,S)-Whelk-O 1 and ChiraSpher. The isomers of 2-butene-1,4-diol can be separated on both chiral columns while the isomers of lafutidine can only be resolved on ChiraSpher column. The influence of different type and amount of mobile phase modifier on the isomers separation was extensively studied. The resolution of cis and trans isomers of 2-butene-1,4-diol was 2.61 on (S,S)-Whelk-O 1 column with hexane-ethanol (97:3, v/v) as the mobile phase. The resolution of lafutidine was 1.89 on ChiraSpher column with hexane-ethanol-THF-diethylamine (92:3:5:0.1, v/v/v/v) as the mobile phase. LC-MS methods were developed to identify the isomer peaks.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B86206
cis-2-Butene-1,4-diol, 97%
C4H8O2