EMAIL THIS PAGE TO A FRIEND
Archives of pharmacal research
Anti-diabetic activities of fucosterol from Pelvetia siliquosa.
PMID 15595413
Abstract
Fucosterol isolated from Pelvetia siliquosa was tested for its anti-diabetic activity in vivo. Fucosterol, when administered orally at 30 mg/kg in streptozotocin-induced diabetic rats, was caused a significant decrease in serum glucose concentrations, and exhibited an inhibition of sorbitol accumulations in the lenses. Fucosterol, when administered orally at 300 mg/kg in epinephrine-induced diabetic rats, was also caused an inhibition of blood glucose level and glycogen degradation. These results demonstrated that fucosterol is a main anti-diabetic principle from the marine algae P. siliquosa.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F5379
Fucosterol, ≥93%
C29H48O