EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Synthesis and pharmacological evaluation of glycine-modified analogues of the neuroprotective agent glycyl-L-prolyl-L-glutamic acid (GPE).
PMID 15598574
Abstract
The synthesis of 10 G*PE analogues, wherein the glycine residue has been modified, is described by coupling readily accessible dibenzyl-L-prolyl-L-glutamate 2 with various analogues of glycine. Pharmacological evaluation of the novel compounds was undertaken to further understand the role of the glycine residue on the observed neuroprotective properties of the endogenous tripeptide GPE.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G3923
Gly-Pro-Glu, ≥98% (HPLC)
C12H19N3O6