Cellular signalling
The distinct stage-specific effects of 2-(p-amylcinnamoyl)amino-4-chlorobenzoic acid on the activation of MAP kinase and Cdc2 kinase in Xenopus oocyte maturation.
PMID 15601628
2-(p-amylcinnamoyl)amino-4-chlorobenzoic acid (PACA), pharmacological inhibitor of phospholipase A(2) (PLA(2)), inhibits epinephrine-stimulated thromboxane production in human platelets. In this study, we investigated the effect of PACA on meiotic maturation individually in stages V and VI oocytes. PACA prevented the maturation in stage V but merely delayed the process in stage VI oocytes. This was associated with the strong inhibition of Mos synthesis at both stages. Besides, PACA-induced inhibition of MAPK activation was evident in stage V but not in stage VI oocytes. PACA also inhibited the activation of Cdc2 kinase (Cdc2) in stage V but merely delayed the process in stage VI oocytes. Furthermore, 5 microM and higher concentrations of PACA completely inhibited the activation of MAPK and Cdc2 only in stage V, not in stage VI, oocytes. Moreover, we propose PACA as a new tool for the study of Xenopus oocyte maturation, which can also play a unique role for the studies of the stage-specific activation of MAPK and Cdc2.
Related Materials
Product #
Molecular Formula
Add to Cart
O0766 ONO-RS-082, ≥97% (HPLC)