EMAIL THIS PAGE TO A FRIEND
Journal of bacteriology
Localization of MreB in Rhodobacter sphaeroides under conditions causing changes in cell shape and membrane structure.
PMID 15601688
Abstract
MreB is thought to be a bacterial actin homolog that defines the morphology of rod-shaped bacteria. Rhodobacter sphaeroides changes shape, from a rod to coccobacillus, and undergoes extensive cytoplasmic membrane invagination when it switches from aerobic to photoheterotrophic growth. The role of MreB in defining R. sphaeroides shape was therefore investigated. Attempts at deleting or insertionally inactivating mreB were unsuccessful under all growth conditions. Immunofluorescence microscopy showed MreB localized to mid-cell in elongating cells under both aerobic and photoheterotrophic conditions. Three-dimensional reconstruction showed that MreB formed a ring at mid-cell. MreB remained at mid-cell as septation began but localized to new sites in the daughter cells before the completion of septation. MreB localized to putative septation sites in cephalexin-treated filamentous cells. Genomic single-copy mreB was replaced with gfp-mreB, and green fluorescent protein (GFP)-MreB localized in the same pattern, as seen with immunofluorescence microscopy. Some of the cells expressing GFP-MreB were abnormal, principally displaying an increase in cell width, suggesting that the fusion was not fully functional in all cells. GFP-MreB localized to swellings at mid-cell in cells treated with the penicillin-binding protein 2 inhibitor amdinocillin. These data suggest that MreB is essential in R. sphaeroides, performing a role at mid-cell in elongating cells, and in early septation, putatively in the cytoplasmic control of the peptidoglycan synthetic complexes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33447
Mecillinam, VETRANAL, analytical standard
C15H23N3O3S