EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Detection of single base mismatches and abasic sites using phenanthridinium as an artificial DNA base and charge donor.
PMID 15602596
Abstract
Combining the fluorescence properties of phenanthridinium as an artificial DNA base together with DNA-mediated charge transfer processes allows the homogeneous detection of DNA base mismatches and abasic sites.