EMAIL THIS PAGE TO A FRIEND
Molecular diversity
Synthesis and dynamic NMR study of ketenimines derived from tert-butyl isocyanide, alkyl 2-arylamino-2-oxo-acetates, and dialkyl acetylenedicarboxylates.
PMID 15612648
Abstract
The adduct produced in the reaction between tert-butyl isocyanide and dialkyl acetylenedicarboxylates was trapped by alkyl 2-arylamino-2-oxo-acetates. When the aryl group is 2-methyl-6-nitrophenyl or 2,6-di-isopropylphenyl, the product exists as two stable rotamers at room temperature as a result of restricted rotation around the Ar-N single bond. When the aryl group is 1-naphthyl or 8-quinolinyl, dynamic NMR effects are observed in the 1H NMR spectra. The calculated free-energy of activation for interconversion of the rotational isomers in 1-naphthyl and 8-quinolinyl derivatives amounts to about 99+/-2 and 68.5+/-2 kJ mol(-1), respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
260630
tert-Butyl isocyanide, 98%
C5H9N