EMAIL THIS PAGE TO A FRIEND
Chinese journal of digestive diseases
Experimental studies of oxymatrine and its mechanisms of action in hepatitis B and C viral infections.
PMID 15612666
Abstract
Viral hepatitis B is a worldwide public health problem; in China, chronic hepatitis B patients account for 30-40 million people and chronic hepatitis C accounts for 16% of all chronic hepatitis patients. Interferon-alpha (IFNalpha) is rather expensive and has many serious side-effects, which makes practitioners prudent in its use. A multicenter study held in China showed that the negative seroconversion of HBV DNA by oxymatrine and IFNalpha-1b was 42.3% and 40.7%, respectively, and that of HBeAg was 36.5% and 38.9%, respectively, after 3 months treatment with each in patients with chronic hepatitis. Oxymatrine has a similar efficacy to IFNalpha, but with no adverse effects apart from slight injection pain, and it is much less expensive.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O0891
Oxymatrine
C15H24N2O2