EMAIL THIS PAGE TO A FRIEND
Nature
Accurate multiplex gene synthesis from programmable DNA microchips.
PMID 15616567
Abstract
Testing the many hypotheses from genomics and systems biology experiments demands accurate and cost-effective gene and genome synthesis. Here we describe a microchip-based technology for multiplex gene synthesis. Pools of thousands of 'construction' oligonucleotides and tagged complementary 'selection' oligonucleotides are synthesized on photo-programmable microfluidic chips, released, amplified and selected by hybridization to reduce synthesis errors ninefold. A one-step polymerase assembly multiplexing reaction assembles these into multiple genes. This technology enabled us to synthesize all 21 genes that encode the proteins of the Escherichia coli 30S ribosomal subunit, and to optimize their translation efficiency in vitro through alteration of codon bias. This is a significant step towards the synthesis of ribosomes in vitro and should have utility for synthetic biology in general.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
11418009001
DNA Molecular Weight Marker IV