EMAIL THIS PAGE TO A FRIEND
Journal of fluorescence
Optical sensor for the determination of glucose based on KIO4 chemiluminescence detection.
PMID 15617266
Abstract
A method to determine glucose using an optical sensor prepared by entrapping glucose oxidase into silica sol-gel column has been developed. The silica sol-gel film was coated on alumina substrate. The optical sensor is based on the chemiluminescence intensity from the reaction of periodate and hydrogen peroxide in K2CO3 medium. The effect of the ratio of water and alcohol for the preparation of TEOS sol on chemiluminescence intensity was investigated. The effects of pH of enzyme reactor, concentrations of potassium periodate and SDS, and flow rate on the chemiluminescence intensity were studied to find the optimum experimental conditions to determine glucose. The chemiluminescence intensity increased linearly with increasing glucose concentration from 5.0 x 10(-4) M to 1.0 x 10(-7) M and the detection limit was 4.0 x 10(-8) M. Interference effects from some metal ions on chemiluminescence intensity were also investigated.