EMAIL THIS PAGE TO A FRIEND
Biology of Reproduction
Factors regulating the timing of puberty onset in female rhesus monkeys (Macaca mulatta): role of prenatal androgens, social rank, and adolescent body weight.
PMID 15625235
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D5879
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
C2H6OS