EMAIL THIS PAGE TO A FRIEND
Biology of reproduction
Expression of alpha5 integrin (Itga5) is elevated in the rat myometrium during late pregnancy and labor: implications for development of a mechanical syncytium.
PMID 15635129
Abstract
The underlying mechanisms controlling uterine contractions during labor are still poorly understood. Integrins are heterodimeric, transmembrane receptors composed of alpha and beta subunits that can be found in focal adhesions. Because these structures play an important role in the regulation of smooth muscle contractility and cell adhesion, we hypothesized that alpha5 integrin mRNA (Itga5) and protein (ITGA5) expression would be induced in the rat myometrium during late pregnancy and labor. Itga5 mRNA expression was significantly increased (P < 0.05) from Day 17 to labor, noticeably decreasing 1 day postpartum (PP). Immunoblot analysis illustrated a continual increase in ITGA5 levels during pregnancy, labor, and PP, with levels reaching significance at labor (P < 0.05). Analysis of ITGA5 expression by immunocytochemistry demonstrated that it is primarily localized to myometrial cell membranes in the longitudinal muscle layer of the myometrium from before pregnancy to Day 6, and in both the longitudinal and circular muscle layers from Day 15 to PP. Treatment of late-pregnant rats with progesterone blocked labor and resulted in sustained expression of Itga5 mRNA expression to Day 24. In addition, immunocytochemistry experiments showed ITGA5 was detectable at higher levels in cell membranes of both myometrial layers in progesterone-treated animals on Days 23 and 24, compared with vehicle controls. We propose that ITGA5, with its sole known partner, ITGB1, may be important in promoting cellular cohesion during late pregnancy. This process may aid the development of a mechanical syncytium for efficient force transduction during the sustained, coordinated, and powerful contractions of labor.