EMAIL THIS PAGE TO A FRIEND
Chemistry and physics of lipids
Investigating the structural and dynamic properties of n-doxylstearic acid in magnetically-aligned phospholipid bilayers by X-band EPR spectroscopy.
PMID 15642584
Abstract
X-band electron paramagnetic resonance (EPR) spectroscopy has been employed to investigate the dynamic properties of magnetically-aligned phospholipid bilayers (bicelles) based on the molecular order parameters (S(mol)), the hyperfine splitting values and the line shapes of the EPR spectra. For the first time, a series of EPR spectra of n-doxylstearic acid spin-labels (n = 5, 7, 12, and 16) incorporated into Tm3+-doped parallel-aligned, Dy3+-doped perpendicular-aligned, and randomly dispersed 1,2-dimyristoyl-sn-glycero-3-phosphocholine/1,2-dihexanoyl-sn-glycero-3-phosphocholine (DMPC/DHPC) bicelles with respect to the direction of the static magnetic field have been investigated as a function of cholesterol content and temperature variation to characterize the orientational aspects along the hydrocarbon acyl chains. Important general observations are that under conditions for which the bicelle is poised in the liquid crystalline phase, the degree of ordering decreases as the nitroxide moiety is transferred toward the end of the stearic acid acyl chains. The addition of cholesterol increases the phase transition temperature and alignment temperature of the DMPC/DHPC phospholipid bilayers and increases the chain order. However, increasing the temperature of the bicelle system decreases the chain order. This report reveals that the dynamic properties of DMPC/DHPC bicelles agree well with other biological and model membrane systems. The results indicate that magnetically-aligned phospholipid bilayers are an excellent model membrane system.