EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Transnitrosation of nitrosothiols: characterization of an elusive intermediate.
PMID 15643848
Abstract
We report an investigation of the reaction between (S)-nitroso-l-cysteine ethyl ester and l-cysteine ethyl ester as a model of physiologically relevant transnitrosation processes. Our theoretical and experimental evidence clearly supports the existence of a nitroxyl disulfide intermediate in solution.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C121908
L-Cysteine ethyl ester hydrochloride, 98%
C5H11NO2S · HCl