EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Evidence against the hopping mechanism as an important electron transfer pathway for conformationally constrained oligopeptides.
PMID 15643851
Abstract
The rate constant of intramolecular electron transfer through oligopeptides based on the alpha-aminoisobutyric acid residue was determined as a function of the peptide length and found to depend weakly on the donor-acceptor separation. By measuring the electron-transfer activation energy and estimating the energy gap between donor and bridge, we were able to discard the electron hopping mechanism.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C89803
4-Cyanobenzoic acid, 99%
C8H5NO2