EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the USA
Melatonin synthesis: 14-3-3-dependent activation and inhibition of arylalkylamine N-acetyltransferase mediated by phosphoserine-205.
PMID 15644438
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB5467 Anti-AANAT Antibody, serum, clone, Chemicon®