EMAIL THIS PAGE TO A FRIEND
Rapid communications in mass spectrometry : RCM
The inactivation of phospholipase A2 by scalaradial: a biomimetic study by electrospray mass spectrometry.
PMID 15645482
Abstract
A biomimetic approach was employed to shed light on the nature of chemical reactions occurring in the covalent inactivation of phospholipase A(2) (PLA(2)) by scalaradial (1), a marine dialdehyde terpenoid endowed with potent anti-inflammatory activity. To this end, a detailed study of the reaction profile between the nitrogenous nucleophile isopropylamine and scalaradial was performed under biologically relevant conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
471291
Isopropylamine, ≥99.5%
C3H9N
45945
Isopropylamine, anhydrous, analytical standard
C3H9N