EMAIL THIS PAGE TO A FRIEND
Journal of neurophysiology
Varicosity-Schwann cell interactions mediated by ATP in the mouse vas deferens.
PMID 15647402
Abstract
Schwann cells, from a variety of sources, are known to possess P2Y purinergic metabotropic receptors. However, it is not known if Schwann cells associated with autonomic nerve terminals possess such receptors and if so whether these receptors are activated by the endogenous release of ATP from the nerve terminals. We show that such Schwann cells in the vas deferens give evoked calcium transients on nerve stimulation. These transients are mediated, at least in part, by the endogenous release of ATP, which acts on Schwann cell P2Y receptors to release calcium from within the cells. This work suggests the possibility that Schwann cells are active participants in the process of junctional transmission in the autonomic nervous system.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C199
CGS-15943, solid
C13H8ClN5O