EMAIL THIS PAGE TO A FRIEND
Cell and Tissue Research
Colocalization of GH, TSH and prolactin, but not ACTH, with betaLH-immunoreactivity: evidence for pluripotential cells in the ovine pituitary.
PMID 15647919