EMAIL THIS PAGE TO A FRIEND
Journal (Canadian Dental Association)
Foil backing used in intraoral radiographic dental film: a source of environmental lead.
PMID 15649339
Abstract
The lead content of the foil backing of 4 types of intraoral film commonly used by dentists was 69% to 85%. An environmental issue exists because these foils are typically thrown out with regular refuse, even though recycling programs exist. For a new adult patient, a full-mouth radiographic series would generate 11.2 g of waste lead; for a 6-month checkup, waste lead would only be produced if radiographs were required. In an experiment that simulated the acidic conditions that might be expected in a landfill site, 3.5-4.4 mg of lead was released during 17-hour incubation in dilute acetic acid. When distilled double-deionized water replaced the acid, 1.0-2.2 mg of lead was released by the same types of foils. Human health concerns also exist when dental assistants handle lead foil while developing radiographs and fail to change their gloves or wash their hands before handling instruments and dental paraphernalia used in the mouth. Although the amount of lead introduced into the oral cavity would be relatively small, the elimination of sources of lead exposure, especially for children, is important.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF96512597
Lead, foil, 150x150mm, thickness 0.5mm, as rolled, 99.95%
Pb
GF74259793
Lead, foil, 150x150mm, thickness 1.0mm, as rolled, 99.5%
Pb
GF57430638
Lead, foil, 150x150mm, thickness 1.0mm, as rolled, 99.95%
Pb
GF63249419
Lead, foil, 150x150mm, thickness 2.0mm, as rolled, 99.95%
Pb
GF60047760
Lead, foil, 250x250mm, thickness 3.0mm, as rolled, 99.95%
Pb
GF05521160
Lead, foil, 25x25mm, thickness 0.125mm, as rolled, 99.95%
Pb
GF42741637
Lead, foil, 25x25mm, thickness 0.125mm, as rolled, 99.99+%
Pb
GF60380940
Lead, foil, 25x25mm, thickness 0.15mm, as rolled, 99.95%
Pb
GF85562994
Lead, foil, 25x25mm, thickness 0.1mm, as rolled, 99.95%
Pb
GF74435519
Lead, foil, 25x25mm, thickness 0.25mm, as rolled, 99.95%
Pb
GF10987757
Lead, foil, 25x25mm, thickness 0.25mm, as rolled, 99.99+%
Pb
GF03985724
Lead, foil, 25x25mm, thickness 0.55mm, as rolled, 99.999%
Pb
GF73081852
Lead, foil, 25x25mm, thickness 0.5mm, as rolled, 99.95%
Pb
GF68898188
Lead, foil, 25x25mm, thickness 0.5mm, as rolled, 99.99+%
Pb
GF90025129
Lead, foil, 25x25mm, thickness 1.0mm, as rolled, 99.5%
Pb
GF55764041
Lead, foil, 25x25mm, thickness 1.0mm, as rolled, 99.95%
Pb
GF85349346
Lead, foil, 25x25mm, thickness 1.0mm, as rolled, 99.99+%
Pb
GF70374810
Lead, foil, 25x25mm, thickness 1.5mm, as rolled, 99.99+%
Pb
GF22900766
Lead, foil, 25x25mm, thickness 3.0mm, as rolled, 99.99+%
Pb
GF36533505
Lead, foil, 300x300mm, thickness 0.125mm, as rolled, 99.95%
Pb
GF00434843
Lead, foil, 300x300mm, thickness 0.15mm, as rolled, 99.95%
Pb
GF87346806
Lead, foil, 300x300mm, thickness 0.1mm, as rolled, 99.95%
Pb
GF50662773
Lead, foil, 300x300mm, thickness 0.25mm, as rolled, 99.95%
Pb
GF98077898
Lead, foil, 1000x1000mm, thickness 0.5mm, as rolled, 99.95%
Pb
GF68213538
Lead, foil, 100x100mm, thickness 0.125mm, as rolled, 99.95%
Pb
GF32778322
Lead, foil, 100x100mm, thickness 0.15mm, as rolled, 99.95%
Pb
GF21778397
Lead, foil, 100x100mm, thickness 0.1mm, as rolled, 99.95%
Pb
GF54864041
Lead, foil, 100x100mm, thickness 0.25mm, as rolled, 99.95%
Pb
GF64074034
Lead, foil, 300x300mm, thickness 0.5mm, as rolled, 99.95%
Pb
GF06345479
Lead, foil, 300x300mm, thickness 1.0mm, as rolled, 99.5%
Pb