EMAIL THIS PAGE TO A FRIEND
The Journal of antimicrobial chemotherapy
Pyridine N-oxide derivatives: unusual anti-HIV compounds with multiple mechanisms of antiviral action.
PMID 15650002
Abstract
Pyridine N-oxide derivatives represent a new class of anti-HIV compounds, for which some members exclusively act through inhibition of HIV-1 reverse transcriptase and thus characteristically behave as non-nucleoside reverse transcriptase inhibitors. Other members act, additionally or alternatively, at a post-integrational event in the replication cycle of HIV, that is, at the level of HIV gene expression. Repeated administration of one of the prototype compounds (JPL-32) to DBA/2 and hu-PBMC-SCID mice demonstrated, in the absence of any acute toxicity, protective activity against HIV-induced destruction of CD4 human T lymphocytes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
131652
Pyridine N-oxide, 95%
C5H5NO