EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Preparation of 8-(arylsulfanyl)adenines with diazonium salts under mild, aerobic conditions.
PMID 15651828
Abstract
8-(Arylsulfanyl)adenines 11 were prepared in up to 75% yield by reacting the 8-thionoadenine 6 (acetic acid 3-(6-amino-8-thioxo-7,8-dihydropurin-9-yl)propyl ester) with benzenediazonium tetrafluoroborates in DMSO. Benzenediazonium ions carrying an electron-withdrawing substituent gave the highest yields. The reaction proceeded smoothly at room temperature without any base and could be performed under air atmosphere. The extremely mild conditions are compatible with a wide range of functional groups.