EMAIL THIS PAGE TO A FRIEND
Schizophrenia research
A missense polymorphism (H204R) of a Rho GTPase-activating protein, the chimerin 2 gene, is associated with schizophrenia in men.
PMID 15653288
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV51862
Anti-CHN2 antibody produced in rabbit, affinity isolated antibody