EMAIL THIS PAGE TO A FRIEND
Skinmed
Seborrheic dermatitis.
PMID 15654167
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
257710
Selenium sulfide
S2Se