EMAIL THIS PAGE TO A FRIEND
Radiobiologiia
[The mechanism of the radioprotective action of sulfolane].
PMID 1565766
Abstract
As a result of radiolysis of sulfolane, that has a radioprotective action, sulfonyl anion-radicals and hydrosulfonyl radicals are produced, subsequent transformations of which lead to complete regeneration of sulfolane. On the other hand, some other five-membered cyclic sulfones, that have no radioprotective action, are irreversibly depleted in reactions with water radiolysis products. Sulfolane is suggested to protect from the effect of e(-)aq and H. and to be ineffective in protecting against OH. radicals.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
88965
Sulfolane, analytical standard
C4H8O2S
T22209
Sulfolane, 99%
C4H8O2S